missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylpropylamine, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 314690050

 View more versions of this product

Catalog No. AC314690050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Methylpropylamine
627-35-0
100.0
0.7100g/mL
<−34°C
95% min. (GC)
C4H11N
CH3CH2CH2NHCH3
5 g
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
CCCNC
73.14
73.14
Liquid
96%
95.0
Colorless
61.0°C to 63.0°C
Authentic
Glass bottle
1.3880 to 1.3910
MFCD00009361
0.71
GVWISOJSERXQBM-UHFFFAOYSA-N
N-methylpropan-1-amine
12315
96%

Chemical Identifiers

627-35-0
73.14
GVWISOJSERXQBM-UHFFFAOYSA-N
12315
CCCNC
C4H11N
MFCD00009361
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
N-methylpropan-1-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only