missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpropylamine, 96%

CAS: 627-35-0 | C4H11N | 73.14 g/mol

$113.35 - $113.35

Chemical Identifiers

CAS 627-35-0
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00009361
InChI Key GVWISOJSERXQBM-UHFFFAOYSA-N
Synonym n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
PubChem CID 12315
IUPAC Name N-methylpropan-1-amine
SMILES CCCNC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC314690050
SDS View Documents
Thermo Scientific Chemicals
314690050
5 g Glass bottle
Each for $113.35
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

627-35-0
73.14
GVWISOJSERXQBM-UHFFFAOYSA-N
12315
CCCNC
C4H11N
MFCD00009361
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
N-methylpropan-1-amine

Specifications

627-35-0
100.0
0.7100g/mL
<−34°C
95% min. (GC)
C4H11N
CH3CH2CH2NHCH3
5 g
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
CCCNC
73.14
73.14
Liquid
95.0
Colorless
61.0°C to 63.0°C
Authentic
Glass bottle
1.3880 to 1.3910
MFCD00009361
0.71
GVWISOJSERXQBM-UHFFFAOYSA-N
N-methylpropan-1-amine
12315
96%
N-Methylpropylamine, 96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only