missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(n-Butyl)aniline, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2418022

 View more versions of this product

Catalog No. AAB2418022


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-(n-Butyl)aniline
0.932
105°C (222°F)
1.532
100 g
benzenamine, n-butyl, aniline, n-butyl, 4-phenylamino butane, n-butylbenzenamine, n-n-butyl aniline, butylaniline, n-phenyl-n-butylamine, butylphenylamine, unii-r2znj7l2um, benzamine, n-butyl
CCCCNC1=CC=CC=C1
149.237
149.23
1126-78-9
240°C to 242°C
C10H15N
MFCD00035798
UN2738
VSHTWPWTCXQLQN-UHFFFAOYSA-N
N-butylaniline
14310
99%

Chemical Identifiers

1126-78-9
149.237
VSHTWPWTCXQLQN-UHFFFAOYSA-N
14310
CCCCNC1=CC=CC=C1
C10H15N
MFCD00035798
benzenamine, n-butyl, aniline, n-butyl, 4-phenylamino butane, n-butylbenzenamine, n-n-butyl aniline, butylaniline, n-phenyl-n-butylamine, butylphenylamine, unii-r2znj7l2um, benzamine, n-butyl
N-butylaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N-BUTYLANILINE

EINECSNumber : 214-425-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only