missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals

N,N-Dicyclohexylcarbodiimide, 99%, C13H22N2, CAS Number-538-75-0, carbodiimide, dicyclohexyl, bis cyclohexyl carbodiimide, dcc, 1,3-dicyclohexylcarbodiimide, carbodicyclohexylimide, n,n'-dicyclohexylcarbodiimide, n,n-dicyclohexylcarbodiimide, dcci, dicyclohexylcarbodiimide, dccd, 1kg

Supplier:  Thermo Scientific Chemicals 113900010

 View more versions of this product

Catalog No. AC113900010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Specifications

N,N′-Dicyclohexylcarbodiimide
98.5
33.0°C to 35.0°C
122.0°C to 124.0°C (6.0 mmHg)
Authentic
Glass bottle
C6H11N=C=NC6H11
1 kg
15,3107
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents,100g/l dichloromethane
C1CCC(CC1)N=C=NC2CCCCC2
206.33
5.81 mPa.s (55°C)
206.33
Crystalline Mass or Waxy Solid
538-75-0
100.0
White to Yellow
>110°C
98.5% min. (GC)
C13H22N2
MFCD00011659
01,231; 02,126; 03,91; 04,141; 05,206; 06,174; 07,100; 08,162; 09,156; 10,142; 11,173; 13,333; 14,131; 15,131; 16,128
dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl
QOSSAOTZNIDXMA-UHFFFAOYSA-N
N,N'-dicyclohexylmethanediimine
10868
CHEBI:53090
99%

Chemical Identifiers

538-75-0
206.33
QOSSAOTZNIDXMA-UHFFFAOYSA-N
10868
N,N'-dicyclohexylmethanediimine
C13H22N2
MFCD00011659
dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl
CHEBI:53090
C1CCC(CC1)N=C=NC2CCCCC2
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 208-704-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only