Learn More
N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N-Dicyclohexylcarbodiimide, 99%, C13H22N2, CAS Number-538-75-0, carbodiimide, dicyclohexyl, bis cyclohexyl carbodiimide, dcc, 1,3-dicyclohexylcarbodiimide, carbodicyclohexylimide, n,n'-dicyclohexylcarbodiimide, n,n-dicyclohexylcarbodiimide, dcci, dicyclohexylcarbodiimide, dccd, 1kg
$116.15 - $1906.22
Chemical Identifiers
CAS | 538-75-0 |
---|---|
Molecular Formula | C13H22N2 |
Molecular Weight (g/mol) | 206.33 |
MDL Number | MFCD00011659 |
InChI Key | QOSSAOTZNIDXMA-UHFFFAOYSA-N |
Synonym | dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl |
PubChem CID | 10868 |
ChEBI | CHEBI:53090 |
IUPAC Name | N,N'-dicyclohexylmethanediimine |
SMILES | C1CCC(CC1)N=C=NC2CCCCC2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC113901000
|
Thermo Scientific Chemicals
113901000 |
100 g | Glass bottle |
Each for $116.15
|
|
||||
AC113902500
|
Thermo Scientific Chemicals
113902500 |
250 g | Glass bottle |
Each for $298.48
|
|
||||
AC113900010
|
Thermo Scientific Chemicals
113900010 |
1 kg | Glass bottle |
Each for $483.90
|
|
||||
AC113900050
|
Thermo Scientific Chemicals
113900050 |
5 kg | Plastic drum |
Each for $1,906.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
538-75-0 | |
206.33 | |
QOSSAOTZNIDXMA-UHFFFAOYSA-N | |
10868 | |
N,N'-dicyclohexylmethanediimine |
C13H22N2 | |
MFCD00011659 | |
dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl | |
CHEBI:53090 | |
C1CCC(CC1)N=C=NC2CCCCC2 |
Specifications
538-75-0 | |
100.0 | |
White to Yellow | |
>110°C | |
98.5% min. (GC) | |
C13H22N2 | |
MFCD00011659 | |
01,231; 02,126; 03,91; 04,141; 05,206; 06,174; 07,100; 08,162; 09,156; 10,142; 11,173; 13,333; 14,131; 15,131; 16,128 | |
dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl | |
QOSSAOTZNIDXMA-UHFFFAOYSA-N | |
N,N'-dicyclohexylmethanediimine | |
10868 | |
CHEBI:53090 | |
99% | |
N,N′-Dicyclohexylcarbodiimide |
98.5 | |
33.0°C to 35.0°C | |
122.0°C to 124.0°C (6.0 mmHg) | |
Authentic | |
Glass bottle | |
C6H11N=C=NC6H11 | |
100 g | |
15,3107 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents,100g/l dichloromethane | |
C1CCC(CC1)N=C=NC2CCCCC2 | |
206.33 | |
5.81 mPa.s (55°C) | |
206.33 | |
Crystalline Mass or Waxy Solid |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 208-704-1
RUO – Research Use Only