Learn More
N,N-Diethylmethylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 114461000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N, N-Diethylmethylamine | |
616-39-7 | |
100.0 | |
Colorless to Yellow | |
63°C to 65°C | |
Authentic | |
Glass bottle | |
1.3890 to 1.391 | |
MFCD00009049 | |
04, 99 | |
n,n-diethylmethylamine, diethylmethylamine, ethanamine, n-ethyl-n-methyl, n-methyldiethylamine, diethylamine, n-methyl, n-ethyl-n-methyl-ethanamine, methyldiethylamine, diethylmethyl amine, diethyl methyl amine, methyl diethyl amine | |
GNVRJGIVDSQCOP-UHFFFAOYSA-O | |
N-ethyl-N-methylethanamine | |
12022 | |
98% |
98% | |
97.5 | |
-196°C | |
0.7200g/mL | |
-23°C | |
97.5% min. (GC) | |
C5H14N | |
(C2H5)2NCH3 | |
100 mL | |
0.72 | |
Solubility in water: soluble. Other solubilities: very soluble in ethanol and ethyl ether,soluble in most common organic solvents | |
CC[NH+](C)CC | |
88.17 | |
87.15 | |
Liquid |
Chemical Identifiers
616-39-7 | |
88.17 | |
GNVRJGIVDSQCOP-UHFFFAOYSA-O | |
12022 | |
CC[NH+](C)CC |
C5H14N | |
MFCD00009049 | |
n,n-diethylmethylamine, diethylmethylamine, ethanamine, n-ethyl-n-methyl, n-methyldiethylamine, diethylamine, n-methyl, n-ethyl-n-methyl-ethanamine, methyldiethylamine, diethylmethyl amine, diethyl methyl amine, methyl diethyl amine | |
N-ethyl-N-methylethanamine |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 210-480-5
TSCA : TSCA
RUO – Research Use Only