missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Diethylmethylamine, 98%, Thermo Scientific Chemicals

$107.22 - $107.22

Chemical Identifiers

CAS 616-39-7
Molecular Formula C5H14N
Molecular Weight (g/mol) 88.17
MDL Number MFCD00009049
InChI Key GNVRJGIVDSQCOP-UHFFFAOYSA-O
Synonym n,n-diethylmethylamine, diethylmethylamine, ethanamine, n-ethyl-n-methyl, n-methyldiethylamine, diethylamine, n-methyl, n-ethyl-n-methyl-ethanamine, methyldiethylamine, diethylmethyl amine, diethyl methyl amine, methyl diethyl amine
PubChem CID 12022
IUPAC Name N-ethyl-N-methylethanamine
SMILES CC[NH+](C)CC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC114461000
SDS View Documents
Thermo Scientific Chemicals
114461000
100 mL Glass bottle
Each for $107.22
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

616-39-7
88.17
GNVRJGIVDSQCOP-UHFFFAOYSA-O
12022
CC[NH+](C)CC
C5H14N
MFCD00009049
n,n-diethylmethylamine, diethylmethylamine, ethanamine, n-ethyl-n-methyl, n-methyldiethylamine, diethylamine, n-methyl, n-ethyl-n-methyl-ethanamine, methyldiethylamine, diethylmethyl amine, diethyl methyl amine, methyl diethyl amine
N-ethyl-N-methylethanamine

Specifications

616-39-7
100.0
Colorless to Yellow
63°C to 65°C
Authentic
Glass bottle
1.3890 to 1.391
MFCD00009049
04, 99
n,n-diethylmethylamine, diethylmethylamine, ethanamine, n-ethyl-n-methyl, n-methyldiethylamine, diethylamine, n-methyl, n-ethyl-n-methyl-ethanamine, methyldiethylamine, diethylmethyl amine, diethyl methyl amine, methyl diethyl amine
GNVRJGIVDSQCOP-UHFFFAOYSA-O
N-ethyl-N-methylethanamine
12022
98%
N, N-Diethylmethylamine, 98%
97.5
-196°C
0.7200g/mL
-23°C
97.5% min. (GC)
C5H14N
(C2H5)2NCH3
100 mL
0.72
Solubility in water: soluble. Other solubilities: very soluble in ethanol and ethyl ether,soluble in most common organic solvents
CC[NH+](C)CC
88.17
87.15
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 210-480-5

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only