missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Diisopropylethylamine, 99.5+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 367840025

 View more versions of this product

Catalog No. AC367840025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N, N-Diisopropylethylamine
7087-68-5
100.0
Colorless to Yellow
127.0°C
Authentic
Glass bottle
1.4123 to 1.4143
MFCD00008868
04,IV,511
0.756
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
JGFZNNIVVJXRND-UHFFFAOYSA-N
N-ethyl-N-propan-2-ylpropan-2-amine
81531
129.24
Liquid
99.5+%
99.5
-46.0°C
0.7560g/mL
6°C
99.5% min. (GC)
C8H19N
[(CH3)2CH]2NC2H5
2.5 L
01,371; 04,230; 07,148; 10,151; 12,277; 13,121; 16,176
15,3219
Solubility in water: 3.9g/L (20°C)
CCN(C(C)C)C(C)C
129.24
0.88 mm2/s (20°C)
99.5+%

Chemical Identifiers

7087-68-5
129.24
JGFZNNIVVJXRND-UHFFFAOYSA-N
81531
CCN(C(C)C)C(C)C
C8H19N
MFCD00008868
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
N-ethyl-N-propan-2-ylpropan-2-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 230-392-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only