missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylamine, 99.5+%

CAS: 7087-68-5 | C8H19N | 129.24 g/mol

$91.60 - $1052.72

Chemical Identifiers

CAS 7087-68-5
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.24
MDL Number MFCD00008868
InChI Key JGFZNNIVVJXRND-UHFFFAOYSA-N
Synonym n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
PubChem CID 81531
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
SMILES CCN(C(C)C)C(C)C
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367840250
SDS View Documents
Thermo Scientific Chemicals
367840250
25 mL Glass bottle
Each for $91.60
Only null left
 
AC367840500
SDS View Documents
Thermo Scientific Chemicals
367840500
50 mL Glass bottle
Each for $141.11
Only null left
 
AC367841000
SDS View Documents
Thermo Scientific Chemicals
367841000
100 mL Glass Bottle
Each for $178.86
Only null left
 
AC367840010
SDS View Documents
Thermo Scientific Chemicals
367840010
1 L Glass Bottle
Each for $854.19
Only null left
 
AC367840025
SDS View Documents
Thermo Scientific Chemicals
367840025
2.5 L Glass bottle
Each for $1,052.72
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7087-68-5
129.24
JGFZNNIVVJXRND-UHFFFAOYSA-N
81531
CCN(C(C)C)C(C)C
C8H19N
MFCD00008868
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
N-ethyl-N-propan-2-ylpropan-2-amine

Specifications

7087-68-5
100.0
Colorless to Yellow
127.0°C
Authentic
Glass bottle
1.4123 to 1.4143
MFCD00008868
04,IV,511
0.756
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
JGFZNNIVVJXRND-UHFFFAOYSA-N
N-ethyl-N-propan-2-ylpropan-2-amine
81531
129.24
Liquid
99.5
-46.0°C
0.7560g/mL
6°C
99.5% min. (GC)
C8H19N
[(CH3)2CH]2NC2H5
25 mL
01,371; 04,230; 07,148; 10,151; 12,277; 13,121; 16,176
15,3219
Solubility in water: 3.9g/L (20°C)
CCN(C(C)C)C(C)C
129.24
0.88 mm2/s (20°C)
99.5+%
N, N-Diisopropylethylamine, 99.5+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 230-392-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only