Learn More
N,N-Diisopropylethylamine, 99.5+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 367841000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N, N-Diisopropylethylamine | |
7087-68-5 | |
100.0 | |
Colorless to Yellow | |
127.0°C | |
Authentic | |
Glass Bottle | |
1.4123 to 1.4143 | |
MFCD00008868 | |
2733 | |
01,371; 04,230; 07,148; 10,151; 12,277; 13,121; 16,176 | |
15,3219 | |
Solubility in water: 3.9g/L (20°C) | |
CCN(C(C)C)C(C)C | |
129.24 | |
0.88 mm2/s (20°C) | |
99.5+% |
99.5+% | |
99.5 | |
-46.0°C | |
0.7560g/mL | |
6°C | |
99.5% min. (GC) | |
C8H19N | |
[(CH3)2CH]2NC2H5 | |
100 mL | |
04,IV,511 | |
0.756 | |
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine | |
JGFZNNIVVJXRND-UHFFFAOYSA-N | |
N-ethyl-N-propan-2-ylpropan-2-amine | |
81531 | |
129.24 | |
Liquid |
Chemical Identifiers
7087-68-5 | |
129.24 | |
JGFZNNIVVJXRND-UHFFFAOYSA-N | |
81531 | |
CCN(C(C)C)C(C)C |
C8H19N | |
MFCD00008868 | |
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine | |
N-ethyl-N-propan-2-ylpropan-2-amine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 230-392-
RUO – Research Use Only