missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Dimethyl-n-octadecylamine 90.0+%, TCI America™
Supplier: TCI America D1609250G
Specifications
N,N-Dimethyl-n-octadecylamine | |
White | |
MFCD00048496 | |
3259 | |
PFKRTWCFCOUBHS-UHFFFAOYSA-N | |
dimethyl(octadecyl)azanium chloride | |
15365 | |
≥90.0% (GC) |
124-28-7 | |
C20H44ClN | |
250 g | |
n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine | |
[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C | |
334.03 | |
297.57 | |
Crystalline Lumps |
Chemical Identifiers
124-28-7 | |
334.03 | |
PFKRTWCFCOUBHS-UHFFFAOYSA-N | |
15365 | |
[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C |
C20H44ClN | |
MFCD00048496 | |
n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine | |
dimethyl(octadecyl)azanium chloride |
Safety and Handling
EINECSNumber : (2)-0176&(2)-0185
RTECSNumber : RG4200000
TSCA : Yes