missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-n-octadecylamine 90.0+%, TCI America™

$72.70 - $165.90

Chemical Identifiers

CAS 124-28-7
Molecular Formula C20H44ClN
Molecular Weight (g/mol) 334.03
MDL Number MFCD00048496
InChI Key PFKRTWCFCOUBHS-UHFFFAOYSA-N
Synonym n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
PubChem CID 15365
IUPAC Name dimethyl(octadecyl)azanium chloride
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
D160925G
View Documents
TCI America
D160925G
25 g
Each for $72.70
Only null left
 
D1609250G
View Documents
TCI America
D1609250G
250 g
Each for $165.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

124-28-7
334.03
PFKRTWCFCOUBHS-UHFFFAOYSA-N
15365
[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
C20H44ClN
MFCD00048496
n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
dimethyl(octadecyl)azanium chloride

Specifications

124-28-7
C20H44ClN
25 g
n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
334.03
297.57
Crystalline Lumps
White
MFCD00048496
3259
PFKRTWCFCOUBHS-UHFFFAOYSA-N
dimethyl(octadecyl)azanium chloride
15365
≥90.0% (GC)
N,N-Dimethyl-n-octadecylamine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0176&(2)-0185

RTECSNumber : RG4200000

TSCA : Yes

SDS
Documents

Documents