missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethyl-p-phenylenediamine oxalate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 408490250

 View more versions of this product

Catalog No. AC408490250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N, N-Dimethyl-p-phenylenediamine oxalate
62778-12-5
100.0
Beige to Brown or Gray
99%
0·5 C2H2O4
25 g
362.42
Chunks and/or Powder
99%
98.5
200.0°C
Authentic
Glass bottle
MFCD00036387
362.42
99%

Chemical Identifiers

62778-12-5
0·5 C2H2O4
362.42
MFCD00036387
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Rinse mouth.
IF IN EYES: Rinse cautiously with water for several minutes.

GHS Signal Word: Danger

EINECSNumber : 263-723-2

RTECSNumber : CZ1586550

TSCA : TSCA

Recommended Storage : this product darkens readily on exposure to air

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only