Learn More
N,N-Dimethyl-p-phenylenediamine oxalate, 99%, Thermo Scientific Chemicals
$166.74 - $676.95
Chemical Identifiers
CAS | 62778-12-5 |
---|---|
Molecular Formula | 0·5 C2H2O4 |
Molecular Weight (g/mol) | 362.42 |
MDL Number | MFCD00036387 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC408490050
|
Thermo Scientific Chemicals
408490050 |
5 g | Glass bottle |
Each for $166.74
|
|
||||
AC408490250
|
Thermo Scientific Chemicals
408490250 |
25 g | Glass bottle |
Each for $381.25
|
|
||||
AC408491000
|
Thermo Scientific Chemicals
408491000 |
100 g | Glass bottle |
Each for $676.95
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62778-12-5 | |
0·5 C2H2O4 | |
362.42 | |
MFCD00036387 |
Specifications
62778-12-5 | |
100.0 | |
Beige to Brown or Gray | |
99% | |
0·5 C2H2O4 | |
5 g | |
362.42 | |
Chunks and/or Powder |
98.5 | |
200.0°C | |
Authentic | |
Glass bottle | |
MFCD00036387 | |
362.42 | |
99% | |
N, N-Dimethyl-p-phenylenediamine oxalate, 99% |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Rinse mouth.
IF IN EYES: Rinse cautiously with water for several minutes.
GHS Signal Word: Danger
EINECSNumber : 263-723-2
RTECSNumber : CZ1586550
TSCA : TSCA
Recommended Storage : this product darkens readily on exposure to air
RUO – Research Use Only