missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N-Dimethylhexanamide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2456222

 View more versions of this product

Catalog No. AAB2456222


Explore available promotions

Description

Description

N,N-Dimethylhexanamide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylhexanamide is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

N,N-Dimethylhexanamide
0.89
C8H17NO
MFCD00043671
1747074
Not miscible or difficult to mix.
CCCCCC(=O)N(C)C
143.23
143.23
5830-30-8
80°C (1 mmHg)
1.449
100 g
hexanamide, n,n-dimethyl, n,n-dimethylcaproamide, n,n-dimethylhexanoamide, nn-dimethylhexanamide, nn-dimethylcaproamide, n,n-dirnethylhexanamide, n,n-dimethyl-hexanamide, hexanamide,n,n-dimethyl, acmc-1atq3, n,n-dimethylhexanamide, tech
OAERLTPBKQBWHJ-UHFFFAOYSA-N
N,N-dimethylhexanamide
22084
98%

Chemical Identifiers

5830-30-8
143.23
OAERLTPBKQBWHJ-UHFFFAOYSA-N
22084
CCCCCC(=O)N(C)C
C8H17NO
MFCD00043671
hexanamide, n,n-dimethyl, n,n-dimethylcaproamide, n,n-dimethylhexanoamide, nn-dimethylhexanamide, nn-dimethylcaproamide, n,n-dirnethylhexanamide, n,n-dimethyl-hexanamide, hexanamide,n,n-dimethyl, acmc-1atq3, n,n-dimethylhexanamide, tech
N,N-dimethylhexanamide
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 227-406-2

RTECSNumber : MN7950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only