missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N-Dimethylhexanamide, 98%, Thermo Scientific Chemicals
$250.67 - $757.21
Chemical Identifiers
CAS | 5830-30-8 |
---|---|
Molecular Formula | C8H17NO |
Molecular Weight (g/mol) | 143.23 |
MDL Number | MFCD00043671 |
InChI Key | OAERLTPBKQBWHJ-UHFFFAOYSA-N |
Synonym | hexanamide, n,n-dimethyl, n,n-dimethylcaproamide, n,n-dimethylhexanoamide, nn-dimethylhexanamide, nn-dimethylcaproamide, n,n-dirnethylhexanamide, n,n-dimethyl-hexanamide, hexanamide,n,n-dimethyl, acmc-1atq3, n,n-dimethylhexanamide, tech |
PubChem CID | 22084 |
IUPAC Name | N,N-dimethylhexanamide |
SMILES | CCCCCC(=O)N(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2456214
|
Thermo Scientific Chemicals
B2456214 |
25 g |
Each for $250.67
|
|
|||||
AAB2456222
|
Thermo Scientific Chemicals
B2456222 |
100 g |
Each for $757.21
|
|
|||||
Description
N,N-Dimethylhexanamide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethylhexanamide is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
5830-30-8 | |
143.23 | |
OAERLTPBKQBWHJ-UHFFFAOYSA-N | |
22084 | |
CCCCCC(=O)N(C)C |
C8H17NO | |
MFCD00043671 | |
hexanamide, n,n-dimethyl, n,n-dimethylcaproamide, n,n-dimethylhexanoamide, nn-dimethylhexanamide, nn-dimethylcaproamide, n,n-dirnethylhexanamide, n,n-dimethyl-hexanamide, hexanamide,n,n-dimethyl, acmc-1atq3, n,n-dimethylhexanamide, tech | |
N,N-dimethylhexanamide |
Specifications
5830-30-8 | |
80°C (1 mmHg) | |
1.449 | |
25 g | |
hexanamide, n,n-dimethyl, n,n-dimethylcaproamide, n,n-dimethylhexanoamide, nn-dimethylhexanamide, nn-dimethylcaproamide, n,n-dirnethylhexanamide, n,n-dimethyl-hexanamide, hexanamide,n,n-dimethyl, acmc-1atq3, n,n-dimethylhexanamide, tech | |
OAERLTPBKQBWHJ-UHFFFAOYSA-N | |
N,N-dimethylhexanamide | |
22084 | |
98% |
0.89 | |
C8H17NO | |
MFCD00043671 | |
1747074 | |
Not miscible or difficult to mix. | |
CCCCCC(=O)N(C)C | |
143.23 | |
143.23 | |
N,N-Dimethylhexanamide |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 227-406-2
RTECSNumber : MN7950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only