Learn More
N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 147490010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N,N,N',N'-Tetramethyl-1,6-hexanediamine | |
111-18-2 | |
100.0 | |
Colorless to Yellow | |
209.0°C to 210.0°C | |
Authentic | |
Glass bottle | |
1.4350 to 1.4370 | |
MFCD00008339 | |
04, I, 423 | |
n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210 | |
TXXWBTOATXBWDR-UHFFFAOYSA-N | |
N,N,N',N'-tetramethylhexane-1,6-diamine | |
8097 | |
172.31 | |
Liquid |
99% | |
98.5 | |
-46.0°C | |
0.8060g/mL | |
73°C | |
99% | |
C10H24N2 | |
(CH3)2N(CH2)6N(CH3)2 | |
1 L | |
0.806 | |
Solubility in water: miscible. Other solubilities: soluble in ethanol, acetone, dichloromethane and, toluene | |
CN(C)CCCCCCN(C)C | |
172.31 | |
1.72 mPa.s (20°C) | |
99% |
Chemical Identifiers
111-18-2 | |
172.31 | |
TXXWBTOATXBWDR-UHFFFAOYSA-N | |
8097 | |
CN(C)CCCCCCN(C)C |
C10H24N2 | |
MFCD00008339 | |
n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210 | |
N,N,N',N'-tetramethylhexane-1,6-diamine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air a
GHS Signal Word: Danger
EINECSNumber : 203-842-9
RTECSNumber : MO1370000
TSCA : TSCA
RUO – Research Use Only