missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals

$95.97 - $1096.19

Chemical Identifiers

CAS 111-18-2
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.31
MDL Number MFCD00008339
InChI Key TXXWBTOATXBWDR-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210
PubChem CID 8097
IUPAC Name N,N,N',N'-tetramethylhexane-1,6-diamine
SMILES CN(C)CCCCCCN(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC147490250
SDS View Documents
Thermo Scientific Chemicals
147490250
25 mL Glass bottle
Each for $95.97
 
AC147492500
SDS View Documents
Thermo Scientific Chemicals
147492500
250 mL Glass bottle
Each for $485.03
 
AC147490010
SDS View Documents
Thermo Scientific Chemicals
147490010
1 L Glass bottle
Each for $1,096.19
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-18-2
172.31
TXXWBTOATXBWDR-UHFFFAOYSA-N
8097
CN(C)CCCCCCN(C)C
C10H24N2
MFCD00008339
n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210
N,N,N',N'-tetramethylhexane-1,6-diamine

Specifications

111-18-2
100.0
Colorless to Yellow
209.0°C to 210.0°C
Authentic
Glass bottle
1.4350 to 1.4370
MFCD00008339
04, I, 423
n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210
TXXWBTOATXBWDR-UHFFFAOYSA-N
N,N,N',N'-tetramethylhexane-1,6-diamine
8097
172.31
Liquid
98.5
-46.0°C
0.8060g/mL
73°C
99%
C10H24N2
(CH3)2N(CH2)6N(CH3)2
25 mL
0.806
Solubility in water: miscible. Other solubilities: soluble in ethanol, acetone, dichloromethane and, toluene
CN(C)CCCCCCN(C)C
172.31
1.72 mPa.s (20°C)
99%
N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air a

GHS Signal Word: Danger

EINECSNumber : 203-842-9

RTECSNumber : MO1370000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only