missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 147492500

 View more versions of this product

Catalog No. AC147492500

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N,N,N',N'-Tetramethyl-1,6-hexanediamine
111-18-2
100.0
Colorless to Yellow
209.0°C to 210.0°C
Authentic
Glass bottle
1.4350 to 1.4370
MFCD00008339
04, I, 423
n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210
TXXWBTOATXBWDR-UHFFFAOYSA-N
N,N,N',N'-tetramethylhexane-1,6-diamine
8097
172.31
Liquid
99%
98.5
-46.0°C
0.8060g/mL
73°C
99%
C10H24N2
(CH3)2N(CH2)6N(CH3)2
250 mL
0.806
Solubility in water: miscible. Other solubilities: soluble in ethanol, acetone, dichloromethane and, toluene
CN(C)CCCCCCN(C)C
172.31
1.72 mPa.s (20°C)
99%

Chemical Identifiers

111-18-2
172.31
TXXWBTOATXBWDR-UHFFFAOYSA-N
8097
CN(C)CCCCCCN(C)C
C10H24N2
MFCD00008339
n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210
N,N,N',N'-tetramethylhexane-1,6-diamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air a

GHS Signal Word: Danger

EINECSNumber : 203-842-9

RTECSNumber : MO1370000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only