missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N,N,N'-Trimethylethylenediamine 97.0+%, TCI America™

$94.79 - $266.62

Chemical Identifiers

CAS 142-25-6
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.181
MDL Number MFCD00014874
InChI Key HVOYZOQVDYHUPF-UHFFFAOYSA-N
Synonym n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine
PubChem CID 67338
IUPAC Name N,N',N'-trimethylethane-1,2-diamine
SMILES CNCCN(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T07425ML
View Documents
TCI America
T07425ML
5 mL
Each for $94.79
 
T074225ML
View Documents
TCI America
T074225ML
25 mL
Each for $266.62
 
Description

Description

Specifications

Chemical Identifiers

142-25-6
102.181
HVOYZOQVDYHUPF-UHFFFAOYSA-N
67338
CNCCN(C)C
C5H14N2
MFCD00014874
n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine
N,N',N'-trimethylethane-1,2-diamine

Specifications

142-25-6
116°C
MFCD00014874
2733
HVOYZOQVDYHUPF-UHFFFAOYSA-N
N,N',N'-trimethylethane-1,2-diamine
67338
≥97.0% (GC,T)
N,N,N′-Trimethylethylenediamine
Colorless
C5H14N2
5 mL
n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine
CNCCN(C)C
102.181
102.18
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'