missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,N,N'-Trimethylethylenediamine 97.0+%, TCI America™
Supplier: TCI America T074225ML
Specifications
N,N,N′-Trimethylethylenediamine | |
Colorless | |
C5H14N2 | |
25 mL | |
n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine | |
CNCCN(C)C | |
102.181 | |
102.18 | |
Liquid |
142-25-6 | |
116°C | |
MFCD00014874 | |
2733 | |
HVOYZOQVDYHUPF-UHFFFAOYSA-N | |
N,N',N'-trimethylethane-1,2-diamine | |
67338 | |
≥97.0% (GC,T) |
Chemical Identifiers
142-25-6 | |
102.181 | |
HVOYZOQVDYHUPF-UHFFFAOYSA-N | |
67338 | |
CNCCN(C)C |
C5H14N2 | |
MFCD00014874 | |
n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine | |
N,N',N'-trimethylethane-1,2-diamine |
Safety and Handling
TSCA : Yes