Learn More
n-Nonane, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 129110025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Nonane | |
98.5 | |
-53.0°C | |
0.7180g/mL | |
31°C | |
98.5% min. (GC) | |
C9H20 | |
CH3(CH2)7CH3 | |
2.5 L | |
0.718 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, slightly soluble in benzene, acetone, chloroform | |
CCCCCCCCC | |
128.26 | |
CHEBI:32892 | |
99% | |
Liquid |
111-84-2 | |
100.0 | |
Colorless | |
151.0°C | |
Authentic | |
Glass bottle | |
1.4040 to 1.4060 | |
MFCD00009574 | |
01, 165 | |
n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard | |
BKIMMITUMNQMOS-UHFFFAOYSA-N | |
nonane | |
8141 | |
128.26 | |
Pure |
Chemical Identifiers
111-84-2 | |
BKIMMITUMNQMOS-UHFFFAOYSA-N | |
8141 | |
nonane |
MFCD00009574 | |
n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard | |
CHEBI:32892 | |
CCCCCCCCC |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 203-913-4
RUO – Research Use Only