missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Nonane, 99%, pure, Thermo Scientific Chemicals

$226.07 - $2567.31

Chemical Identifiers

CAS 111-84-2
MDL Number MFCD00009574
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC129111000
SDS View Documents
Thermo Scientific Chemicals
129111000
100 mL Glass bottle
Each for $226.07
 
AC129115000
SDS View Documents
Thermo Scientific Chemicals
129115000EA
500 mL Glass bottle
Each for $730.97
 
AC129110025
SDS View Documents
Thermo Scientific Chemicals
129110025
2.5 L Glass bottle
Each for $2,567.31
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-84-2
BKIMMITUMNQMOS-UHFFFAOYSA-N
8141
nonane
MFCD00009574
n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
CHEBI:32892
CCCCCCCCC

Specifications

111-84-2
100.0
Colorless
151.0°C
Authentic
Glass bottle
1.4040 to 1.4060
MFCD00009574
01, 165
n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
BKIMMITUMNQMOS-UHFFFAOYSA-N
nonane
8141
128.26
Pure
n-Nonane
98.5
-53.0°C
0.7180g/mL
31°C
98.5% min. (GC)
C9H20
CH3(CH2)7CH3
100 mL
0.718
Solubility in water: insoluble. Other solubilities: soluble in alcohol, slightly soluble in benzene, acetone, chloroform
CCCCCCCCC
128.26
CHEBI:32892
99%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 203-913-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only