Learn More
n-Octyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1596422
Description
n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in alcohol, phenoxy ethanol and propylene glycol. Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
n-Octyl 4-hydroxybenzoate | |
50°C to 53°C | |
MFCD00016482 | |
octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german | |
RIKCMEDSBFQFAL-UHFFFAOYSA-N | |
octyl 4-hydroxybenzoate | |
14642 | |
98% |
1219-38-1 | |
C15H22O3 | |
100 g | |
Soluble in alcohol,phenoxy ethanol and propylene glycol. Insoluble in water. | |
CCCCCCCCOC(=O)C1=CC=C(O)C=C1 | |
250.34 | |
250.34 |
Chemical Identifiers
1219-38-1 | |
250.34 | |
RIKCMEDSBFQFAL-UHFFFAOYSA-N | |
14642 | |
CCCCCCCCOC(=O)C1=CC=C(O)C=C1 |
C15H22O3 | |
MFCD00016482 | |
octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german | |
octyl 4-hydroxybenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-943-2
RTECSNumber : DH2529050
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only