missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Octyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

$76.38 - $702.11

Chemical Identifiers

CAS 1219-38-1
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00016482
InChI Key RIKCMEDSBFQFAL-UHFFFAOYSA-N
Synonym octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german
PubChem CID 14642
IUPAC Name octyl 4-hydroxybenzoate
SMILES CCCCCCCCOC(=O)C1=CC=C(O)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1596406
View Documents
Thermo Scientific Chemicals
A1596406
5 g
Each for $76.38
 
AAA1596414
View Documents
Thermo Scientific Chemicals
A1596414
25 g
Each for $238.27
 
AAA1596422
View Documents
Thermo Scientific Chemicals
A1596422
100 g
Each for $702.11
 
Description

Description

n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in alcohol, phenoxy ethanol and propylene glycol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1219-38-1
250.34
RIKCMEDSBFQFAL-UHFFFAOYSA-N
14642
CCCCCCCCOC(=O)C1=CC=C(O)C=C1
C15H22O3
MFCD00016482
octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german
octyl 4-hydroxybenzoate

Specifications

1219-38-1
C15H22O3
5 g
Soluble in alcohol,phenoxy ethanol and propylene glycol. Insoluble in water.
CCCCCCCCOC(=O)C1=CC=C(O)C=C1
250.34
250.34
n-Octyl 4-hydroxybenzoate
50°C to 53°C
MFCD00016482
octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german
RIKCMEDSBFQFAL-UHFFFAOYSA-N
octyl 4-hydroxybenzoate
14642
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-943-2

RTECSNumber : DH2529050

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only