missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Octyl-β-D-glucopyranoside (White Crystalline Powder), Fisher BioReagents
Supplier: Fisher BioReagents FLBP5855
Description
n-Octyl-β-D-Glucopyranoside is a highly purified, nonionic detergent.Specifications
0.3 max. at 260nm | |
White | |
Powder/Solid | |
5 g | |
≥98 % | |
C14H28O6 | |
15, 6857 | |
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O | |
292.37 | |
292.4 |
n-Octyl-β-D-Glucopyranoside | |
29836-26-8 | |
105°C | |
Amber Poly Bottle | |
-28.0 to -32.0° (+ or -) | |
MFCD00063288 | |
HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol | |
62852 |
Chemical Identifiers
29836-26-8 | |
292.37 | |
HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O |
C14H28O6 | |
MFCD00063288 | |
62852 |
Safety and Handling
Emergency Overview
The toxicological properties have not been fully investigated.
NFPA
Health: 1
Flammability:
Signal Word: CAUTION!
Recommended Storage : -20°C