missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Octyl-β-D-glucopyranoside (White Crystalline Powder), Fisher BioReagents

$771.05 - $2590.00

Chemical Identifiers

CAS 29836-26-8
Molecular Formula C14H28O6
Molecular Weight (g/mol) 292.37
MDL Number MFCD00063288
InChI Key HEGSGKPQLMEBJL-RKQHYHRCSA-N
PubChem CID 62852
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP585-5
SDS View Documents
Fisher BioReagents
FLBP5855
5 g Amber Poly Bottle
Each for $771.05
Case of 6 Each for $4,626.30
 
BP585-25
SDS View Documents
Fisher BioReagents
FLBP58525
25 g Amber Poly Bottle
Each for $2,590.00
 
Description

Description

n-Octyl-β-D-Glucopyranoside is a highly purified, nonionic detergent.
Specifications

Chemical Identifiers

29836-26-8
292.37
HEGSGKPQLMEBJL-RKQHYHRCSA-N
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
C14H28O6
MFCD00063288
62852

Specifications

0.3 max. at 260nm
29836-26-8
105°C
Amber Poly Bottle
-28.0 to -32.0° (+ or -)
MFCD00063288
HEGSGKPQLMEBJL-RKQHYHRCSA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol
62852
n-Octyl-β-D-Glucopyranoside
White
Powder/Solid
5 g
≥98 %
C14H28O6
15, 6857
CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
292.37
292.4
Safety and Handling

Safety and Handling

Emergency Overview
The toxicological properties have not been fully investigated.
NFPA
Health: 1
Flammability:

Signal Word: CAUTION!

Recommended Storage : -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'