missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Pentane For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™
For gas chromatography ECD and FID SupraSolv™
Supplier: Technidata 1008821000
Specifications
| n-Pentane | |
| 109-66-0 | |
| 0.63g/cm3 at 20°C | |
| 1 L | |
| 285°C (545°F) | |
| Glass Bottle | |
| C5H12 | |
| 570 hPa (20°C) | |
| n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
| OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
| pentane | |
| 8003 | |
| 72.15 |
| -130°C | |
| Colorless | |
| 36°C | |
| 48°C (54°F) | |
| Benzine-like | |
| Liquid | |
| MFCD00009498 | |
| 1265 | |
| Soluble in water: 0.4g/L at 16°C | |
| CCCCC | |
| 72.15 | |
| CHEBI:37830 |
Chemical Identifiers
| 109-66-0 | |
| 72.15 | |
| OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
| 8003 | |
| pentane |
| C5H12 | |
| MFCD00009498 | |
| n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
| CHEBI:37830 | |
| CCCCC |
Safety and Handling
H304: May be fatal if swallowed and enters airways.
H336 May cause drowsiness and dizziness.
H225: Highly flammable liquid and vapour.
Recommended Storage : 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.