missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Pentyl acetate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1085236

 View more versions of this product

Catalog No. AAL1085236


Explore available promotions

Description

Description

n-Pentyl acetate is used as in studies of olfactory function and in studies of the psychosocial effects of odor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Pentyl acetate is used as in studies of olfactory function and in studies of the psychosocial effects of odor.

Solubility
Slightly soluble in water (2g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
Specifications

Specifications

n-Pentyl acetate
-100°C
142°C
Fruit-like
1.403
500 g
1744753
Slightly soluble in water (2g/L).
CCCCCOC(=O)C
130.187
CHEBI:87362
99%
628-63-7
0.876
24°C (75°F)
C7H14O2
MFCD00009500
UN1104
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
PGMYKACGEOXYJE-UHFFFAOYSA-N
pentyl acetate
12348
130.19

Chemical Identifiers

628-63-7
130.187
PGMYKACGEOXYJE-UHFFFAOYSA-N
12348
pentyl acetate
C7H14O2
MFCD00009500
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
CHEBI:87362
CCCCCOC(=O)C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H226-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMYL ACETATES

EINECSNumber : 211-047-3

RTECSNumber : AJ1925000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only