missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Pentyl acetate, 99%, Thermo Scientific Chemicals

$49.48 - $112.14

Chemical Identifiers

CAS 628-63-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009500
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
Synonym amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
PubChem CID 12348
ChEBI CHEBI:87362
IUPAC Name pentyl acetate
SMILES CCCCCOC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1085222
View Documents
Thermo Scientific Chemicals
L1085222
100 g
Each for $49.48
 
AAL1085236
View Documents
Thermo Scientific Chemicals
L1085236
500 g
Each for $112.14
 
Description

Description

n-Pentyl acetate is used as in studies of olfactory function and in studies of the psychosocial effects of odor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Pentyl acetate is used as in studies of olfactory function and in studies of the psychosocial effects of odor.

Solubility
Slightly soluble in water (2g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
Specifications

Chemical Identifiers

628-63-7
130.187
PGMYKACGEOXYJE-UHFFFAOYSA-N
12348
pentyl acetate
C7H14O2
MFCD00009500
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
CHEBI:87362
CCCCCOC(=O)C

Specifications

628-63-7
0.876
24°C (75°F)
C7H14O2
MFCD00009500
UN1104
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
PGMYKACGEOXYJE-UHFFFAOYSA-N
pentyl acetate
12348
130.19
n-Pentyl acetate
-100°C
142°C
Fruit-like
1.403
100 g
1744753
Slightly soluble in water (2g/L).
CCCCCOC(=O)C
130.187
CHEBI:87362
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H226-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMYL ACETATES

EINECSNumber : 211-047-3

RTECSNumber : AJ1925000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only