missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl 7-Methoxycoumarin-3-carboxylate 98.0+%, TCI America™

$62.29 - $612.46

Chemical Identifiers

CAS 150321-92-9
Molecular Formula C15H11NO7
Molecular Weight (g/mol) 317.253
MDL Number MFCD00467593
InChI Key JMQAALOXLOSYCQ-UHFFFAOYSA-N
Synonym 7-Methoxycoumarin-3-carboxylic Acid N-Succinimidyl Ester
PubChem CID 2760554
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 7-methoxy-2-oxochromene-3-carboxylate
SMILES COC1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)ON3C(=O)CCC3=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S0867100MG
View Documents
TCI America
S0867100MG
100 mg
Each for $62.29
 
S08671G
View Documents
TCI America
S08671G
1 g
Each for $612.46
 
Description

Description

Specifications

Chemical Identifiers

150321-92-9
317.253
JMQAALOXLOSYCQ-UHFFFAOYSA-N
2760554
COC1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)ON3C(=O)CCC3=O
C15H11NO7
MFCD00467593
7-Methoxycoumarin-3-carboxylic Acid N-Succinimidyl Ester
(2,5-dioxopyrrolidin-1-yl) 7-methoxy-2-oxochromene-3-carboxylate

Specifications

150321-92-9
C15H11NO7
100 mg
JMQAALOXLOSYCQ-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 7-methoxy-2-oxochromene-3-carboxylate
2760554
≥98.0% (HPLC,N)
N-Succinimidyl 7-Methoxycoumarin-3-carboxylate
White
MFCD00467593
7-Methoxycoumarin-3-carboxylic Acid N-Succinimidyl Ester
COC1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)ON3C(=O)CCC3=O
317.253
317.25
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'