missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Succinimidyl 7-Methoxycoumarin-3-carboxylate 98.0+%, TCI America™

Supplier:  TCI America S0867100MG

 View more versions of this product

Catalog No. S0867100MG


Explore available promotions

Description

Description

Specifications

Specifications

N-Succinimidyl 7-Methoxycoumarin-3-carboxylate
White
MFCD00467593
7-Methoxycoumarin-3-carboxylic Acid N-Succinimidyl Ester
COC1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)ON3C(=O)CCC3=O
317.253
317.25
Crystalline Powder
150321-92-9
C15H11NO7
100 mg
JMQAALOXLOSYCQ-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 7-methoxy-2-oxochromene-3-carboxylate
2760554
≥98.0% (HPLC,N)

Chemical Identifiers

150321-92-9
317.253
JMQAALOXLOSYCQ-UHFFFAOYSA-N
2760554
COC1=CC2=C(C=C1)C=C(C(=O)O2)C(=O)ON3C(=O)CCC3=O
C15H11NO7
MFCD00467593
7-Methoxycoumarin-3-carboxylic Acid N-Succinimidyl Ester
(2,5-dioxopyrrolidin-1-yl) 7-methoxy-2-oxochromene-3-carboxylate
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions