missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
N-(α,α,α-Trifluoro-p-tolyl)piperazine, 98%, Thermo Scientific™
Identifiants chimiques
CAS | 30459-17-7 |
---|---|
Molecular Formula | C11H13F3N3 |
Molecular Weight (g/mol) | 230.23 |
MDL Number | MFCD00040765 |
InChI Key | IBQMAPSJLHRQPE-UHFFFAOYSA-N |
Synonym | 1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy |
PubChem CID | 121718 |
IUPAC Name | 1-[4-(trifluoromethyl)phenyl]piperazine |
SMILES | C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
AC367120010
|
Thermo Scientific Chemicals
367120010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
AC367120050
|
Thermo Scientific Chemicals
367120050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
30459-17-7 | |
230.23 | |
IBQMAPSJLHRQPE-UHFFFAOYSA-N | |
121718 | |
C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F |
C11H13F3N3 | |
MFCD00040765 | |
1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy | |
1-[4-(trifluoromethyl)phenyl]piperazine |
Spécifications
30459-17-7 | |
100.0 | |
97.5% min. (GC) | |
C11H13F3N3 | |
1g | |
IBQMAPSJLHRQPE-UHFFFAOYSA-N | |
1-[4-(trifluoromethyl)phenyl]piperazine | |
121718 | |
98% |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00040765 | |
1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy | |
C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F | |
230.23 | |
230.23 | |
N-(α, α, α-Trifluoro-p-tolyl)piperazine, 98% |
Sécurité et manipulation
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 250-210-3
RUO â Research Use Only