3M™ N95 Healthcare Particulate Respirators and Surgical Masks, 1860 Series
Disposable respirators provide protection against certain airborne biological particles. Fluid resistant to splash and spatter of blood and other infectious material.
Supplier:  3M™ 7000002007
 View more versions of this product
Catalog No. 18-991
Explore available promotions
Description
Description
Disposable respirators provide protection against certain airborne biological particles. Fluid resistant to splash and spatter of blood and other infectious material.
  • Minimizes wearer exposure to airborne particles generated by electrocautery, laser surgery and powered medical instruments
  • Fluid resistant to splash and spatter of blood and other infectious materials
  • >99% BFE (Bacterial Filtration Efficiency) according to ASTM™ F2101
  • Two sizes available to fit a broad range of faces
  • Contains no components made from natural rubber latex
  • Designed for ease of breathing
  • Braided headbands, cushioning nose foam
Specifications
Specifications
N - Not Resistant to Oil
ASTM F2101, ASTM F1862
NIOSH 42 CFR 84, N95, approval number: TC-84A-0006
Cup/Molded
Surgical N95
Healthcare Particulate Respirator and Surgical Mask
No
Yes
1860S
Adjustable Nose Clip
Non-sterile
Yes
10
Health Canada Class I Medical Device
OSHA 1910.134
Single-use
Polypropylene, Polyester
Small
99%
Braided Polyisoprene
Aluminum
Fluid 120 mmHg
Braided and Stapled Headbands