missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ N95 Healthcare Particulate Respirators and Surgical Masks, 1860 Series
Disposable respirators provide protection against certain airborne biological particles. Fluid resistant to splash and spatter of blood and other infectious material.
$53.15 - $53.34
Specifications
| Aerosol Type | N - Not Resistant to Oil |
|---|---|
| Assigned Protection Factor (APF) | 10 |
| Certifications/Compliance | ASTM F2101, ASTM F1862 |
| Certifications (CSA) | Health Canada Class I Medical Device |
| Certifications (NIOSH) | NIOSH 42 CFR 84, N95, approval number: TC-84A-0006 |
Description
Disposable respirators provide protection against certain airborne biological particles. Fluid resistant to splash and spatter of blood and other infectious material.
- Minimizes wearer exposure to airborne particles generated by electrocautery, laser surgery and powered medical instruments
- Fluid resistant to splash and spatter of blood and other infectious materials
- >99% BFE (Bacterial Filtration Efficiency) according to ASTM™ F2101
- Two sizes available to fit a broad range of faces
- Contains no components made from natural rubber latex
- Designed for ease of breathing
- Braided headbands, cushioning nose foam
Specifications
| N - Not Resistant to Oil | |
| ASTM F2101, ASTM F1862 | |
| NIOSH 42 CFR 84, N95, approval number: TC-84A-0006 | |
| Cup/Molded | |
| Surgical N95 | |
| Healthcare Particulate Respirator and Surgical Mask | |
| 99% | |
| Braided Polyisoprene | |
| Adjustable Nose Clip | |
| Non-sterile | |
| Yes |
| 10 | |
| Health Canada Class I Medical Device | |
| OSHA 1910.134 | |
| Single-use | |
| Polypropylene, Polyester | |
| No | |
| Yes | |
| Aluminum | |
| Fluid 120 mmHg | |
| Braided and Stapled Headbands |