missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Sterile Syringe Filters
Filter sample volumes of 10 to 100mL with these sterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
Supplier: Thermo Scientific™ 7232520
Description
Thermo Scientific™ Nalgene™ syringe filters are built to meet your labâs clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.
Key Features- 10 to 100mL sample size
- Inlet: Female Luer-Lok™
- Outlet: Male Luer slip
- Polypropylene housing with ID color rings
- Pore size and membrane type printed on unit
Surfactant-free Cellulose Acetate (SFCA)
- Low protein binding, lower extractables than standard CA
- Good for cell culture fluids and cellular solutions
- Polypropylene housings with blue color rings
Cellulose Acetate (CA)
- Some protein binding and extractables
- For sterile filtration and clarification of: buffers, aqueous solutions and reagents
- Polypropylene housings with red color rings
Nylon
- Naturally hydrophilic, low extractables, some protein binding
- Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
- Polypropylene housings with pink color rings
- Naturally hydrophobic
- Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
- Polypropylene housings with white color rings
Specifications
Membrane Filter | |
0.2 μm | |
Sterilization and clarification of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids | |
50 | |
10 to 100 mL (Sample) |
SFCA | |
Sterile | |
Polypropylene (Housing) | |
Syringe Filter | |
25 mm |