Learn More
Thermo Scientific™ Nalgene™ Sterile Syringe Filters
Filter sample volumes of 10 to 100mL with these sterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
$456.00 - $1581.24
Catalog Number | Mfr. No. | Diameter (Metric) | Material (Membrane) | Pore Size | Sterility | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Material (Membrane) | Pore Size | Sterility | Price | Quantity | ||
09-740-100
|
Thermo Scientific™
7201320 |
13 mm | PES | 0.2 μm | Sterile |
Case of 100 Each for $1,301.82
|
|
||
09-740-104
|
Thermo Scientific™
7222520 |
25 mm | Glass Fibers (GF) + Cellulose Acetate (CA) | 0.2 μm | Sterile |
Case of 50 Each for $835.79
|
|
||
09-740-105
|
Thermo Scientific™
7232520 |
25 mm | SFCA | 0.2 μm | Sterile |
Case of 50 Each for $650.91
|
|
||
09-740-106
|
Thermo Scientific™
7232545 |
25 mm | SFCA | 0.45 μm | Sterile |
Case of 50 Each for $650.91
|
|
||
09-740-107
|
Thermo Scientific™
7232580 |
25 mm | Cellulose Acetate | 0.8 μm | Sterile |
Case of 50 Each for $650.91
|
|
||
09-740-108
|
Thermo Scientific™
7239920 |
25 mm | SFCA | 0.2 μm | Sterile |
Case of 125 Each for $1,581.24
|
|
||
09-740-109
|
Thermo Scientific™
7239945 |
25 mm | SFCA | 0.45 μm | Sterile |
Case of 125 Each for $1,494.53
|
|
||
09-740-113
|
Thermo Scientific™
7252520 |
25 mm | PES | 0.2 μm | Sterile |
Case of 50 Each for $650.91
|
|
||
09-740-114
|
Thermo Scientific™
7252545 |
25 mm | PES | 0.45 μm | Sterile |
Case of 50 Each for $456.00
|
|
||
09-740-115
|
Thermo Scientific™
7262520 |
25 mm | Nylon | 0.2 μm | Sterile |
Case of 50 Each for $789.09
|
|
||
09-740-116
|
Thermo Scientific™
7262545 |
25 mm | Nylon | 0.45 μm | Sterile |
Case of 50 Each for $789.09
|
|
||
09-740-34A
|
Thermo Scientific™
1710020 |
4 mm | Cellulose Acetate | 0.2 μm | Non-sterile |
Pack of 100 Each for $798.94
Case of 4 Pack for $3,195.76
|
|
||
09-740-34B
|
Thermo Scientific™
N1710045 |
4 mm | Cellulose Acetate | 0.45 μm | Non-sterile |
Pack of 100 Each for $817.97
Case of 4 Pack for $3,271.88
|
|
||
Description
Thermo Scientific™ Nalgene™ syringe filters are built to meet your labâs clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.
Key Features- 10 to 100mL sample size
- Inlet: Female Luer-Lok™
- Outlet: Male Luer slip
- Polypropylene housing with ID color rings
- Pore size and membrane type printed on unit
- Highest throughput and lowest clogging
- Low binding
- Most commonly used general purpose membrane
- Low protein binding, lower extractables than standard CA
- Good for cell culture fluids and cellular solutions
- Polypropylene housings with blue color rings
- Some protein binding and extractables
- For sterile filtration and clarification of: buffers, aqueous solutions and reagents
- Polypropylene housings with red color rings
- Naturally hydrophilic, low extractables, some protein binding
- Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
- Polypropylene housings with pink color rings
- Naturally hydrophobic
- Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
- Polypropylene housings with white color rings
Specifications
Syringe Filter |