Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure: Bulk Pack
Store and ship light-sensitive reagents with these opaque, amber, HDPE bottles, ideal for diagnostics, biologicals, specialty chemicals, adhesives and veterinary pharmaceuticals.
Supplier: Thermo Scientific™ 3120850002
Description
Store and ship light-sensitive reagents with Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high-density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
- Injection blow-molded for uniform wall thickness and strength
- Opaque amber HDPE Bottle with opaque amber PP Closure
- Meets USP light transmission standards
- Tough plastic construction
- Bulk packed for packaging applications
- Leakproof†
Recommended for:
Storage and shipping of light-sensitive reagents.
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
2 oz., 60 mL | |
2 oz. | |
Narrow-Mouth Packaging Bottles | |
HDPE | |
1000 | |
20 mm | |
Yes | |
Round | |
Packer Bottle | |
Polypropylene | |
37 mm | |
No |
60 mL | |
Opaque Amber | |
14 mm | |
1000 | |
Opaque Amber | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition | |
Narrow | |
85 mm | |
3.34 in. | |
20-415 mm | |
Screw (Bulk Packed, Unassembled) |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.