Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure: Bulk Pack
Store and ship light-sensitive reagents with these opaque, amber, HDPE bottles, ideal for diagnostics, biologicals, specialty chemicals, adhesives and veterinary pharmaceuticals.
$594.77 - $3114.74
Specifications
Color | Opaque Amber |
---|---|
Description | Narrow-Mouth Packaging Bottles |
Material | HDPE |
Closure Color | Opaque Amber |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
03-313-8A
|
Thermo Scientific™
3120850001 |
30 mL | 20 mm |
Case of 1000 Each for $3,114.74
|
|
||||
03-313-8B
|
Thermo Scientific™
3120850002 |
60 mL | 20-415 mm |
Case of 1000 Each for $3,055.13
|
|
||||
03-313-8C
|
Thermo Scientific™
3120850004 |
125 mL | 24-415 mm |
Case of 500 Each for $1,761.65
|
|
||||
03-313-8D
|
Thermo Scientific™
3120850008 |
250 mL | 24-415 mm |
Case of 250 Each for $1,345.03
|
|
||||
03-313-8E
|
Thermo Scientific™
3120850016 |
500 mL | 28-415 mm |
Case of 125 Each for $951.13
|
|
||||
03-313-8F
|
Thermo Scientific™
3120850032 |
1000 mL | 38-430 mm |
Case of 50 Each for $594.77
|
|
||||
Description
Store and ship light-sensitive reagents with Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high-density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Features:- Opaque amber HDPE Bottle with opaque amber PP Closure
- Tough plastic construction
- Leakproof† when used with Nalgene closures
Recommended for:
Storage and shipping of light-sensitive reagents.
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton.
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance DatabaseSpecifications
Opaque Amber | |
HDPE | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition | |
Narrow | |
Polypropylene | |
No |
Narrow-Mouth Packaging Bottles | |
Opaque Amber | |
Yes | |
Round | |
Screw (Bulk Packed, Unassembled) |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.