missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ PETG Certified Clean Biotainer Bottle
For Use With: Specifically designed for pharmaceutical and biological manufacturing; Closure Material: Polypropylene; Color: Clear
Supplier: Thermo Scientific™ 38341542
Description
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™ Nalgene™ Certified Clean PETG Biotainer™ Bottles reduce the risk of carry-over contamination.
- Specifically designed for pharmaceutical and biological manufacturing
- Certified Clean, validated to USP <788>
- <0.5 EU/mL Endotoxin
Specifications
Not Autoclavable | |
5 L | |
White | |
Polypropylene | |
Clear | |
48 mm | |
299 mm | |
21.375 in. | |
6/Cs. | |
Biotainer | |
14.75 in. | |
6 Bottles |
169.07 fl. oz. | |
USP <778> | |
LDPE | |
48 mm | |
166 mm | |
Media/Buffer Preparation â, Media/Buffer Storage, Sampling, Storage, Transportation â, Validation Testing, Drug Substance, Drug Product | |
Neck Handle | |
PETG | |
1 Ea. | |
Sterile | |
USP <788> (Low particulate) |
For Research Use Only. Not for use in diagnostic procedures.