missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ PETG Certified Clean Biotainer Bottle
For Use With: Specifically designed for pharmaceutical and biological manufacturing; Closure Material: Polypropylene; Color: Clear
$1931.74 - $3742.94
Specifications
Autoclavable | Not Autoclavable |
---|---|
Certifications/Compliance | USP <778> |
Closure Color | White |
Closure Liner | LDPE |
Closure Material | Polypropylene |
Catalog Number | Mfr. No. | Capacity (Metric) | Includes | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Includes | Quantity | Price | Quantity | |||
14388011
|
Thermo Scientific™
38302542 |
125 mL | Side Handle | 20 x 5 Bottles |
Case of 100 Each for $3,456.65
|
|
|||
14388012
|
Thermo Scientific™
38300542 |
500 mL | Side Handle | 14 x 5 Bottles |
Case of 70 Each for $3,742.94
|
|
|||
14388013
|
Thermo Scientific™
38311042 |
1 L | Side Handle | 7 x 5 Bottles |
Case of 35 Each for $3,709.79
|
|
|||
14388015
|
Thermo Scientific™
38323042 |
2 L | Side Handle | 4 x 5 Bottles |
Case of 20 Each for $3,251.71
|
|
|||
14388014
|
Thermo Scientific™
38323020 |
2 L | Side Handle | 20 Bottles |
Case of 20 Each for $3,200.47
|
|
|||
14388016
|
Thermo Scientific™
38341516 |
5 L | Side Handle | 6 Bottles |
Case of 6 Each for $2,001.06
|
|
|||
14388017
|
Thermo Scientific™
38341542 |
5 L | Neck Handle | 6 Bottles |
Case of 6 Each for $1,931.74
|
|
|||
Description
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™ Nalgene™ Certified Clean PETG Biotainer™ Bottles reduce the risk of carry-over contamination.
- Specifically designed for pharmaceutical and biological manufacturing
- Certified Clean, validated to USP <788>
- <0.5 EU/mL Endotoxin
Specifications
Not Autoclavable | |
White | |
Polypropylene | |
Media/Buffer Preparation â, Media/Buffer Storage, Sampling, Storage, Transportation â, Validation Testing, Drug Substance, Drug Product | |
Biotainer | |
USP <788> (Low particulate) |
USP <778> | |
LDPE | |
Clear | |
PETG | |
Sterile |
For Research Use Only. Not for use in diagnostic procedures.