Learn More
Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
Ensure consistently reliable and accurate fluid transfers. This durable high-purity tubing is ideal for pharmaceutical, laboratory and bioprocess manufacturing.
Supplier: Thermo Scientific™ 86000070
Description
Ensure consistently reliable and accurate fluid transfers with Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing for Peristaltic Pumpsis.This flexible, durable high-purity translucent tubing is designed for a variety of pump and transfer applications, including pharmaceutical, laboratory and bioprocess manufacturing.
- Tasteless, odorless, non-cytotoxic and non-hemolytic
- Contains no phthalate plasticizers or peroxides that can leach out into transported fluid
- Low binding suface to minimize absorption of proteins and food products
- Autoclavable; also can be sterilized by gamma radiation, EtO and chemical procedures
- Durometer hardness: Shore A, 50
Ordering Information
Double-bagged in case of 50 ft (15.2m)
Specifications
Flexible Tubing | |
Autoclavable | |
6.4 mm | |
7/16 in. | |
50 ft. | |
2.4 mm | |
50 (Shore A) | |
17 psig at 73°F (23°C) |
Meets USP<87> Cytotoxicity (Agar Diffusion and MEM Elution methods); USP<88> Class VI; USP<661>Physicochemical Tests; ISO10993-3 Bacterial Mutagenicity – AmesAssay; ISO10993-4 Hemolysis Direct Contact, EP 3.1.9 Silicone Elastomer f | |
1/4 in. | |
Platinum-cured Silicone | |
11.1 mm | |
3/32 in. | |
Translucent | |
4 x OD bend radius |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.