missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Polycarbonate Baffled Culture Flasks

Increase mixing efficiency with these polycarbonate flasks, an alternative to glass flasks that produce equivalent growth curves with E. coli and S. cerevisiae.

Supplier:  Thermo Scientific™ 41100500

 View more versions of this product

Catalog No. 10-042-5L



Explore available promotions

Alert:

Repeated autoclaving may result in some loss of mechanical strength.


Description

Description

Increase mixing efficiency using Thermo Scientific™ Nalgene™ Polycarbonate Baffled Culture Flasks, an alternative to glass flasks that produce equivalent growth curves with E. coli and S. cerevisiae. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.

  • Polycarbonate is break-resistant and clear; easy to read graduations help identify liquid level with ease.
  • Bottom baffles provide increased mixing, aeration, and gas exchange.
  • Flasks are excellent for a variety of lab procedures, including fermentation, cell culture grown and harvesting, and generic engineering procedures.
  • 500 and 100mL sizes accept either standard 38mm stainless-steel or plastic closures for versatility.
  • Flasks are autoclavable.
Specifications

Specifications

16.9 oz.
500 mL
12
Autoclavable
Baffled
Flask Baffled Straight Neck PC 500mL
Polycarbonate
4
Yes
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions