Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality HDPE Bottles
Perform countless field applications utilizing these general-purpose, wide-mouth, lab quality HDPE bottles.
Supplier: Thermo Scientific™ 21040001
Description
Utilize Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality HDPE Bottles for countless field applications. These containers exhibit excellent chemical resistance to most acids, bases and alcohols and are suitable for freezer use to -100°C and are Leakproof† when used with Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:- Excellent chemical resistance to most corrosives make these bottles versatile and indispensable in your lab
- Ideal for storing, shipping, and packaging liquids — uniform walls provide durability and resistance to splitting or punctures
- Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
- Participates in My Green Labs' ACT Label program for Accountability, Consistency and Transparency around product sustainability
Includes:
Polypropylene screw closure
Specifications
1 oz., 30 mL | |
1 oz. | |
Lab Quality Wide-Mouth Bottle | |
Storage and Shipping of Small Volume, Light Sensitive, Valuable Reagents | |
72 | |
Non-sterile | |
28 mm | |
Yes | |
Wide | |
Yes | |
Wide Mouth Lab Quality Bottle | |
Unlined | |
28 mm | |
Screw Cap |
30 mL | |
Translucent | |
21 mm | |
HDPE | |
12 | |
Natural | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
Reusable | |
Round | |
63 mm | |
3.89 in. | |
Polypropylene | |
34 mm | |
No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.