Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality HDPE Bottles
Perform countless field applications utilizing these general-purpose, wide-mouth, lab quality HDPE bottles.
$89.29 - $907.55
Specifications
Description Lab Quality Wide-Mouth Bottle
Material HDPE
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Disposable Reusable
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
02-893-5A Edge
View Documents
Thermo Scientific™
21040001
30 mL 28 mm
Pack of 12 Each for $89.29
Case of 6 Pack for $535.74
Edge
02-893-5B Edge
View Documents
Thermo Scientific™
21040002
60 mL 28 mm
Pack of 12 Each for $91.45
Case of 6 Pack for $548.70
Edge
02-893-5C Edge
View Documents
Thermo Scientific™
N21040004
125 mL 38 mm
Pack of 12 Each for $115.87
Case of 6 Pack for $695.22
Edge
02-893-5D
View Documents
Thermo Scientific™
21040008
250 mL 43 mm
Pack of 12 Each for $163.84
Case of 6 Pack for $983.04
02-893-5E Edge
View Documents
Thermo Scientific™
21040016
500 mL 53 mm
Pack of 12 Each for $219.74
Case of 4 Pack for $878.96
Edge
02-893-5F Edge
View Documents
Thermo Scientific™
21040032
1 L 63 mm
Pack of 6 Each for $202.84
Case of 4 Pack for $811.36
Edge
10-346-9
View Documents
Thermo Scientific™
21040048
1500 mL 63 mm
Case of 24 Each for $907.55
Description
Description
Utilize Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality HDPE Bottles for countless field applications. These containers exhibit excellent chemical resistance to most acids, bases and alcohols and are suitable for freezer use to -100°C and are Leakproof† when used with Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:
  • Excellent chemical resistance to most corrosives make these bottles versatile and indispensable in your lab
  • Ideal for storing, shipping, and packaging liquids — uniform walls provide durability and resistance to splitting or punctures
  • Linerless caps provide the ultimate in leakproof† protection without the use ofa liner that can wrinkle, cause leaks, or contaminate your reagents
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Includes:
Polypropylene screw closure
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
Specifications
Specifications
Lab Quality Wide-Mouth Bottle
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Reusable
Wide Mouth Lab Quality Bottle
No
HDPE
Yes
Wide
Screw Cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.