missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nalidixic acid sodium salt, 98.1%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215172405

 View more versions of this product

Catalog No. ICN15172405


Explore available promotions

Description

Description

Specifications

Specifications

Nalidixic Acid Sodium Salt
White
C12H12N2O3
nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O
232.239
CHEBI:100147
Powder
389-08-2
∽ 98%
5 g
MHWLWQUZZRMNGJ-UHFFFAOYSA-N
1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
4421
254.2

Chemical Identifiers

389-08-2
232.239
nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
CHEBI:100147
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O
C12H12N2O3
MHWLWQUZZRMNGJ-UHFFFAOYSA-N
4421
1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
Videos
Safety and Handling

Safety and Handling

storageNote1 : Use only in area provided with appropriate exhaust ventilation

Recommended Storage : Store at 0°C.

SDS
Documents

Documents

Promotions

Promotions