missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nalidixic acid sodium salt, 98.1%, MP Biomedicals™

$65.29 - $115.04

Chemical Identifiers

CAS 389-08-2
Molecular Formula C12H12N2O3
Molecular Weight (g/mol) 232.239
InChI Key MHWLWQUZZRMNGJ-UHFFFAOYSA-N
Synonym nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
PubChem CID 4421
ChEBI CHEBI:100147
IUPAC Name 1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15172401
View Documents
MP Biomedicals Inc
0215172401
1 g
Each for $65.29
 
ICN15172405
View Documents
MP Biomedicals Inc
0215172405
5 g
Each for $115.04
 
Description

Description

Specifications

Chemical Identifiers

389-08-2
232.239
nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
CHEBI:100147
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O
C12H12N2O3
MHWLWQUZZRMNGJ-UHFFFAOYSA-N
4421
1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid

Specifications

389-08-2
∽ 98%
1 g
MHWLWQUZZRMNGJ-UHFFFAOYSA-N
1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
4421
254.2
Nalidixic Acid Sodium Salt
White
C12H12N2O3
nalidixic acid, nalidixin, nevigramon, nalidixate, uronidix, innoxalon, nalidixan, nalitucsan, sicmylon, unaserus
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O
232.239
CHEBI:100147
Powder
Videos
Safety and Handling

Safety and Handling

storageNote1 : Use only in area provided with appropriate exhaust ventilation

Recommended Storage : Store at 0°C.

SDS
missing translation for 'documents'

missing translation for 'documents'