missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-Boc-L-arginine hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6361114
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Nalpha-Boc-L-arginine hydrochloride | |
White | |
MFCD00065552 | |
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, n-boc-l-arginine hydrochloride monohydrate, boc-d-arg-oh hcl h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, n-, pubchem14932, pubchem14932 | |
HDELGKMVZYHPPB-RZUXQYDTNA-N | |
(2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate;hydrochloride | |
2729004 | |
98% | |
Powder |
35897-34-8 | |
C11H23ClN4O4 | |
3658939 | |
Slightly soluble in water. | |
Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O | |
310.78 | |
310.78 | |
25 g |
Chemical Identifiers
35897-34-8 | |
310.78 | |
HDELGKMVZYHPPB-RZUXQYDTNA-N | |
2729004 |
C11H23ClN4O4 | |
MFCD00065552 | |
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, n-boc-l-arginine hydrochloride monohydrate, boc-d-arg-oh hcl h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, n-, pubchem14932, pubchem14932 | |
Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only