missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-Boc-L-arginine hydrochloride, 98%, Thermo Scientific Chemicals
$55.37 - $183.15
Chemical Identifiers
CAS | 35897-34-8 |
---|---|
Molecular Formula | C11H23ClN4O4 |
Molecular Weight (g/mol) | 310.78 |
MDL Number | MFCD00065552 |
InChI Key | HDELGKMVZYHPPB-RZUXQYDTNA-N |
Synonym | boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, n-boc-l-arginine hydrochloride monohydrate, boc-d-arg-oh hcl h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, n-, pubchem14932, pubchem14932 |
PubChem CID | 2729004 |
SMILES | Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6361106
|
Thermo Scientific Chemicals
H6361106 |
5 g |
Each for $55.37
|
|
|||||
AAH6361114
|
Thermo Scientific Chemicals
H6361114 |
25 g |
Each for $183.15
|
|
|||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
35897-34-8 | |
310.78 | |
HDELGKMVZYHPPB-RZUXQYDTNA-N | |
2729004 |
C11H23ClN4O4 | |
MFCD00065552 | |
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, n-boc-l-arginine hydrochloride monohydrate, boc-d-arg-oh hcl h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, n-, pubchem14932, pubchem14932 | |
Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O |
Specifications
35897-34-8 | |
C11H23ClN4O4 | |
3658939 | |
Slightly soluble in water. | |
Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O | |
310.78 | |
310.78 | |
5 g | |
Nalpha-Boc-L-arginine hydrochloride |
White | |
MFCD00065552 | |
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, n-boc-l-arginine hydrochloride monohydrate, boc-d-arg-oh hcl h2o, boc-d-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932 | |
HDELGKMVZYHPPB-RZUXQYDTNA-N | |
(2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate;hydrochloride | |
2729004 | |
98% | |
Powder |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only