missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ NanoDrop™ 2000/2000c Spectrophotometers

New sales of this product have been discontinued. Please see the newest model: Thermo Scientific NanoDrop One Microvolume UV-Vis Spectrophotometer.

Supplier:  Thermo Scientific™ ND2000LAPTOP

 View more versions of this product

Catalog No. 13-400-411

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

New sales of this product have been discontinued. Please see the newest model: Thermo Scientific NanoDrop One Microvolume UV-Vis Spectrophotometer.

The patented sample retention system of the NanoDrop 2000 and 2000c spectrophotometers allows a sample to be pipetted directly onto an optical measurement surface. Once the measurement is complete, the surfaces are simply wiped with a lint-free lab wipe. The NanoDrop 2000c integrates the sample retention system with cuvette capability.

NanoDrop 2000

  • Micro-volume sample size (0.5-2.0μL)
  • Easy to use—pipet sample directly onto the pedestal
  • No dilutions required, even for highly concentrated samples
  • Fast measurement time of less than 5 seconds
  • Full spectral output (190 - 840 nm)
  • Preconfigured methods for A260, A280, labeled DNA or Proteins, and much more
  • User-friendly software that includes custom methods and data export capabilities

NanoDrop 2000c

  • Combines all features of the NanoDrop 2000 with cuvette capability in one instrument
  • Advanced micro-volume pedestal technology
  • Dual modes—choose cuvette or pedestal
  • Broader concentration range for measuring very low or very high concentrations
  • Cuvette capability allows for kinetics (time or time/temperature studies) and cell culture (OD 600) measurements

NanoDrop 2000c - Cuvette

  • Heating: 37°C, ±0.5°C
  • Stirring: 150 - 850rpm
  • Z-Height: 8.5mm
  • Cuvette dimensions: 12.5 x 12.5mm, up to 48mmH
  • Cuvette path length options: 10, 5, 2 and 1mm
  • Type: Masked cuvette
  • Absorbance Range: 0.008 to 1.5
  • Measurement Time: <3 sec.
  • Weight: 2.1kg

Compatible with:

Professional versions of Microsoft™ Windows™ XP (32-bit) w/Service Pack (SP) 2 or later, Vista™ (32-bit), and Windows™ 7 (32 bit and 64 bit)

Specifications

Specifications

3% (at 0.74 Abs at 350 nm)
0.002
Microsoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
USB
2 ng/μL (dsDNA) ng/μL
2048-element Linear Silicon CCD Array
Microvolume
Spectrophotometer
190 to 840 nm
Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification
Laptop Computer
NanoDrop 2000 with laptop computer
0.05, 0.1, 0.2, and 1.0 mm
0.5 to 2.0 μL
Microsoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
1 year parts and labor
±1 nm
14 cm
0 to 300 A
UL/CSA and CE
2 ng/μL to 15,000 ng/μL (dsDNA) ng/μL
20 cm
2 to 15,000 ng/μL (dsDNA), 0.10 - 400mg/mL (BSA)
Xenon Flash
180 to 240 nm
12 VDC
14 x 20 cm
Laptop Computer
<5 sec.
1
12 V DC
1.8 nm (FWHM at Hg 253.7)
0.5 to 2.0 μL
5 W
2 kg
Videos
SDS
Documents

Documents

Promotions

Promotions

US Patents 6,628,382 and 6,809,826.